1-Boc-5-methoxyindole-2-boronic acid

Synonyms: N-Boc-5-methoxy-2-indolylboronic acid, (1-(tert-Butoxycarbonyl)-5-methoxy-1H-indol-2-yl)boronic acid

680516-10G 680516-1G
220088-858EA 263.77 USD
220088-858 220088-859
1-Boc-5-methoxyindole-2-boronic acid
1-Boc-5-methoxyindole-2-boronic acid
Formula: C₁₄H₁₈BNO₅
MW: 291.11 g/mol
MDL Number: MFCD04039006
CAS Number: 290331-71-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR