Mercury(I) nitrate dihydrate, Reagent Grade

Danger

Synonyms: Dimercury dinitrate dihydrate, Mercurous nitrate dihydrate

680265-50G 680265-250G
220088-824EA 71.38 USD
220088-824 220088-823
Mercury(I) nitrate dihydrate, Reagent Grade
Mercury(I) nitrate dihydrate
Formula: Hg₂N₂O₆·2H₂O
MW: 561.22 g/mol
Melting Pt: 70 °C
Density: 4.78 g/cm³ (20 °C)
MDL Number: MFCD00150812
CAS Number: 14836-60-3
EINECS: 233-886-4
UN: 1627
ADR: 6.1,II
Merck Index: 13,05923

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR