2-(Bromomethyl)phenylboronic acid

Synonyms: 2-(Bromomethyl)benzeneboronic acid, Dihydroxy(2-(bromomethyl)phenyl)borane, (2-(Bromomethyl)phenyl)metaboric acid, 2-Bromomethylphenylboronic acid

679453-1G 679453-5G
220087-768EA 21.46 USD
220087-768 220088-703
2-(Bromomethyl)phenylboronic acid
2-(Bromomethyl)phenylboronic acid
Formula: C₇H₈BBrO₂
MW: 214.85 g/mol
MDL Number: MFCD01318941
CAS Number: 91983-14-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR