3-(Bromomethyl)phenylboronic acid ≥90%

Synonyms: 3-Bromomethylphenylboronic acid, 3-(Bromomethyl)benzeneboronic acid, Dihydroxy(3-(bromomethyl)phenyl)borane, (3-(Bromomethyl)phenyl)metaboric acid

679445-5G 679445-1G
220087-767EA 113.54 USD
220087-767 220087-766
3-(Bromomethyl)phenylboronic acid ≥90%
3-(Bromomethyl)phenylboronic acid
Formula: C₇H₈BBrO₂
MW: 214.85 g/mol
MDL Number: MFCD01632207
CAS Number: 51323-43-4
UN: 3261
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR