4-(Bromomethyl)phenylboronic acid

Synonyms: (4-(Bromomethyl)phenyl)metaboric acid, (4-(Bromomethyl)phenyl)boronic acid, Dihydroxy(4-(bromomethyl)phenyl)borane, 4-Bromomethylphenylboronic acid, 4-(Bromomethyl)benzeneboronic acid

679437-5G 679437-1G
220087-765EA 75.46 USD
220087-765 220087-764
4-(Bromomethyl)phenylboronic acid
4-(Bromomethyl)phenylboronic acid
Formula: C₇H₈BBrO₂
MW: 214.85 g/mol
MDL Number: MFCD01318113
CAS Number: 68162-47-0
UN: 3261
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR