3-Methoxypropene ≥97%

Synonyms: Allyl methyl ether

678201-25G 678201-5G
220088-600EA 414.62 USD
220088-600 220088-601
3-Methoxypropene ≥97%
3-Methoxypropene
Formula: C₄H₈O
MW: 72.11 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00008649
CAS Number: 627-40-7
EINECS: 210-997-6
UN: 3271
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR