2-Bromo-3,5-dimethoxytoluene

677736-1G 677736-5G
220088-572EA 98.86 USD
220088-572 220088-573
2-Bromo-3,5-dimethoxytoluene
2-Bromo-3,5-dimethoxytoluene
Formula: C₉H₁₁BrO₂
MW: 231.09 g/mol
CAS Number: 13321-73-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR