2-Amino-4-chloropyridine

Warning

Synonyms: 4-Chloro-2-pyridylamine, 4-Chloropyridin-2-amine

676020-25G 676020-5G
220087-986EA 210 USD
220087-986 220087-987
2-Amino-4-chloropyridine
2-Amino-4-chloropyridine
Formula: C₅H₅ClN₂
MW: 128.56 g/mol
MDL Number: MFCD04113820
CAS Number: 19798-80-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR