1-Boc-indole-2-boronic acid

Synonyms: N-(tert-Butoxycarbonyl)-1H-indole-2-boronic acid, 1H-Indole-2-boronic acid, N-BOC protected, N-Boc-indole-2-boronic Acid, 1-(tert-Butoxycarbonyl)indole-2-boronic acid, N-Boc-indole-2-boronic acid

675911-1G 675911-5G
220087-919EA 39.46 USD
220087-919 220087-920
1-Boc-indole-2-boronic acid
1-Boc-indole-2-boronic acid
Formula: C₁₃H₁₆BNO₄
MW: 261.08 g/mol
MDL Number: MFCD02093045
CAS Number: 213318-44-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR