4-Nitrophenylboronic acid

Synonyms: 4-Nitrobenzeneboronic acid, (4-Nitrophenyl)boronic acid, Dihydroxy(4-nitrophenyl)borane, (4-Nitrophenyl)metaboric acid

673854-5G 673854-1G
220088-215EA 127.38 USD
220088-215 220088-214
4-Nitrophenylboronic acid
4-Nitrophenylboronic acid
Formula: C₆H₆BNO₄
MW: 166.93 g/mol
MDL Number: MFCD00161360
CAS Number: 24067-17-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR