2-Cyano-4-methylpyridine

Synonyms: 4-Methylpyridine-2-carbonitrile, 4-Methylpicolinonitrile, 2-Cyano-γ-picoline, 2-Cyano-4-picoline, 4-Methyl-2-pyridinecarbonitrile

673463-1G 673463-5G
220088-176EA 164.77 USD
220088-176 220088-177
2-Cyano-4-methylpyridine
2-Cyano-4-methylpyridine
Formula: C₇H₆N₂
MW: 118.14 g/mol
Boiling Pt: 145 - 148 °C (38 -rr)
Melting Pt: 90 - 91 °C
MDL Number: MFCD00128868
CAS Number: 1620-76-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR