5-Hydroxynicotinic acid

Synonyms: 5-Hydroxypyridine-3-carboxylic acid, 5-Hydroxy-3-pyridinecarboxylic acid

672742-1G 672742-5G
220088-108EA 649.92 USD
220088-108 220088-109
5-Hydroxynicotinic acid
5-Hydroxynicotinic acid
Formula: C₆H₅NO₃
MW: 139.11 g/mol
MDL Number: MFCD00129117
CAS Number: 27828-71-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR