1-Ethyl-3-methylimidazolium dimethylphosphate

671657-50G 671657-5G
220088-036EA 329 USD
220088-036 220088-037
1-Ethyl-3-methylimidazolium dimethylphosphate
1-Ethyl-3-methylimidazolium dimethylphosphate
Formula: C₈H₁₇N₂O₄P
MW: 236.21 g/mol
CAS Number: 945611-27-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR