1-Ethyl-3-methylimidazolium diethylphosphate

Synonyms: 1-ethyl-3-methylimidazolium diethylphosphate

671541-50G 671541-5G
220087-902EA 285 USD
220087-902 220088-017
1-Ethyl-3-methylimidazolium diethylphosphate
1-Ethyl-3-methylimidazolium diethylphosphate
Formula: C₁₀H₂₁N₂O₄P
MW: 264.26 g/mol
MDL Number: MFCD09953486
CAS Number: 848641-69-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR