ω-Bromo-3-fluoroacetophenone ≥97%

Synonyms: Bromomethyl m-fluorophenyl ketone, 3-Fluorophenacyl bromide, 2-Bromo-1-(3-fluorophenyl)ethan-1-one, α-Bromo-3-fluoroacetophenone, 2-Bromo-3'-fluoroacetophenone

669482-1G 669482-5G
220087-072EA 47.38 USD
220087-072 220087-073
ω-Bromo-3-fluoroacetophenone ≥97%
ω-Bromo-3-fluoroacetophenone
Formula: C₈H₆BrFO
MW: 217.04 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00109756
CAS Number: 53631-18-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR