2-Fluorophenyl methyl sulfone ≥97%

Synonyms: 1-Fluoro-2-(methylsulphonyl)benzene

669385-1G 669385-10G
220087-056EA 400.62 USD
220087-056 220087-055
2-Fluorophenyl methyl sulfone ≥97%
2-Fluorophenyl methyl sulfone
Formula: C₇H₇FO₂S
MW: 174.2 g/mol
MDL Number: MFCD03094199
CAS Number: 654-47-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR