2-Bromo-4-fluoro-6-methylaniline ≥97%

Synonyms: 6-Bromo-4-fluoro-o-toluidine

669369-10G 669369-1G
220087-053EA 219.69 USD
220087-053 220087-054
2-Bromo-4-fluoro-6-methylaniline ≥97%
2-Bromo-4-fluoro-6-methylaniline
Formula: C₇H₇BrFN
MW: 204.04 g/mol
MDL Number: MFCD01861196
CAS Number: 202865-77-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR