2-Chlorophenyl methyl sulfone ≥97%

Synonyms: 1-Chloro-2-(methylsulphonyl)benzene

669342-1G 669342-10G
220087-483EA 649.92 USD
220087-483 220087-482
2-Chlorophenyl methyl sulfone ≥97%
2-Chlorophenyl methyl sulfone
Formula: C₇H₇ClO₂S
MW: 190.65 g/mol
CAS Number: 17482-05-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR