2-Bromophenyl methyl sulfone ≥97%

Synonyms: 2-Bromophenyl Methyl sulphone, 1-Bromo-2-(methylsulphonyl)benzene

669334-10G 669334-1G
220087-480EA 480 USD
220087-480 220087-481
2-Bromophenyl methyl sulfone ≥97%
2-Bromophenyl methyl sulphone
Formula: C₇H₇BrO₂S
MW: 235.1 g/mol
MDL Number: MFCD07368924
CAS Number: 33951-33-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR