4-Iodo-3-nitroaniline ≥97%

667927-5G 667927-1G
220087-332EA 499.85 USD
220087-332 220087-331
4-Iodo-3-nitroaniline ≥97%
4-Iodo-3-nitroaniline
Formula: C₆H₅IN₂O₂
MW: 264.02 g/mol
MDL Number: MFCD08458843
CAS Number: 105752-04-3

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR