2-Chloro-6-methoxybenzothiazole

Synonyms: 2-Chloro-6-methoxy-1,3-benzothiazole, 2-Chloro-6-methoxybenzo[d]thiazole

667331-5G 667331-1G
220086-812EA 362.92 USD
220086-812 220086-811
2-Chloro-6-methoxybenzothiazole
2-Chloro-6-methoxybenzothiazole
Formula: C₈H₆ClNOS
MW: 199.66 g/mol
MDL Number: MFCD00671510
CAS Number: 2605-14-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR