3-Amino-5-bromopyridine

Warning

Synonyms: 5-Bromopyridin-3-amine, 5-Bromo-3-pyridylamine

667129-1G 667129-10G
220087-258EA 42.86 USD
220087-258 220085-816
3-Amino-5-bromopyridine
3-Amino-5-bromopyridine
Formula: C₅H₅BrN₂
MW: 173.01 g/mol
MDL Number: MFCD01646060
CAS Number: 13535-01-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR