3-Bromo-N,N-dimethylaniline ≥97%

666939-5G 666939-1G
220087-244EA 98.65 USD
220087-244 220087-243
3-Bromo-N,N-dimethylaniline ≥97%
3-Bromo-N,N-dimethylaniline
Formula: C₈H₁₀BrN
MW: 200.08 g/mol
Boiling Pt: 70 °C (0.4 mmHg)
Melting Pt: 11 °C
Density: 1.3652 g/cm³ (19 °C)
MDL Number: MFCD00045020
CAS Number: 16518-62-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR