4-Bromo-2-furaldehyde

Synonyms: 4-Bromofurfural, 4-Bromofuran-2-carbaldehyde

666599-1G 666599-250MG
220087-196EA 103.17 USD
220087-196 220087-197
4-Bromo-2-furaldehyde
4-Bromo-2-furaldehyde
Formula: C₅H₃BrO₂
MW: 174.98 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00671504
CAS Number: 21921-76-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR