3-Amino-5-tert-butylisoxazole

666580-25G 666580-5G
220087-194EA 358.85 USD
220087-194 220087-195
3-Amino-5-tert-butylisoxazole
3-Amino-5-tert-butylisoxazole
Formula: C₇H₁₂N₂O
MW: 140.19 g/mol
MDL Number: MFCD00055620
CAS Number: 55809-36-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR