2-Bromo-5-pyridineboronic acid

Synonyms: 6-Bromo-3-pyridinylboronic acid, 2-Bromopyridine-5-boronic acid, (6-Bromopyridin-3-yl)boronic acid, 2-Bromo-5-pyridinylboronic acid, 6-Bromopyridine-3-boronic acid

666556-1G 666556-5G
220087-188EA 32.05 USD
220087-188 220087-189
2-Bromo-5-pyridineboronic acid
2-Bromo-5-pyridineboronic acid
Formula: C₅H₅BBrNO₂
MW: 201.81 g/mol
MDL Number: MFCD03411558
CAS Number: 223463-14-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR