7-Methyladenine

Synonyms: 7-Methyl-7H-purin-6-amine

666548-500MG 666548-100MG
220087-187EA 637.54 USD
220087-187 220087-186
7-Methyladenine
7-Methyladenine
Formula: C₆H₇N₅
MW: 149.16 g/mol
MDL Number: MFCD00127790
CAS Number: 935-69-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR