2-Chloro-4-pyridineboronic acid

Synonyms: (2-Chloropyridin-4-yl)boronic acid, 2-Chloro-4-pyridinylboronic acid, 2-Chloropyridine-4-boronic acid

666513-1G 666513-5G
220087-181EA 24.92 USD
220087-181 220087-182
2-Chloro-4-pyridineboronic acid
2-Chloro-4-pyridineboronic acid
Formula: C₅H₅BClNO₂
MW: 157.36 g/mol
MDL Number: MFCD03788416
CAS Number: 458532-96-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR