2-Chloroimidazole

Synonyms: 2-Chloro-1H-imidazole

666408-1G 666408-250MG
220087-169EA 222.92 USD
220087-169 220087-170
2-Chloroimidazole
2-Chloroimidazole
Formula: C₃H₃ClN₂
MW: 102.52 g/mol
MDL Number: MFCD02179530
CAS Number: 16265-04-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR