2,4-Dibromo-N-methylaniline ≥97%

665886-5G 665886-1G
220086-075EA 473.08 USD
220086-075 220086-074
2,4-Dibromo-N-methylaniline ≥97%
2,4-Dibromo-N-methylaniline
Formula: C₇H₇Br₂N
MDL Number: MFCD08276870
CAS Number: 73557-58-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR