2-Hydroxy-3-nitro-6-picoline

Synonyms: 6-Methyl-3-nitro-2(1H)-pyridinone, 2-Hydroxy-6-methyl-3-nitropyridine, 6-Hydroxy-5-nitro-2-picoline, 6-Methyl-3-nitropyridin-2-ol, 6-Methyl-3-nitro-2-pyridone, 2-Methyl-5-nitro-6-pyridone, 6-Hydroxy-2-methyl-5-nitropyridine

665835-1G 665835-5G
220087-140EA 312.92 USD
220087-140 220087-141
2-Hydroxy-3-nitro-6-picoline
2-Hydroxy-3-nitro-6-picoline
Formula: C₆H₆N₂O₃
MW: 154.13 g/mol
MDL Number: MFCD03001414
CAS Number: 39745-39-6
EINECS: 244-647-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR