2-Bromo-5-fluorobenzyl alcohol ≥97%

Synonyms: (2-Bromo-5-fluorophenyl)methanol

665827-1G 665827-5G
220087-138EA 312.92 USD
220087-138 220087-139
2-Bromo-5-fluorobenzyl alcohol ≥97%
2-Bromo-5-fluorobenzyl alcohol
Formula: C₇H₆BrFO
MW: 205.03 g/mol
MDL Number: MFCD00142876
CAS Number: 202865-66-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR