2,3-Bis(trifluoromethyl)pyridine

665657-250MG 665657-1G
220087-120EA 108.46 USD
220087-120 220087-119
2,3-Bis(trifluoromethyl)pyridine
2,3-Bis(trifluoromethyl)pyridine
Formula: C₇H₃F₆N
MW: 215.1 g/mol
Boiling Pt: 158 °C (1013 hPa)
Melting Pt: –2 °C
MDL Number: MFCD03093763
CAS Number: 1644-68-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR