2,6-Bis(trifluoromethyl)pyridine

665649-1G 665649-250MG
220087-117EA 143.23 USD
220087-117 220087-118
2,6-Bis(trifluoromethyl)pyridine
2,6-Bis(trifluoromethyl)pyridine
Formula: C₇H₃F₆N
MW: 215.1 g/mol
MDL Number: MFCD00236675
CAS Number: 455-00-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR