4-Bromo-o-toluonitrile ≥97%

Synonyms: 4-Bromo-2-methylbenzonitrile

665320-5G 665320-25G
220086-962EA 45.12 USD
220086-962 220086-410
4-Bromo-o-toluonitrile ≥97%
4-Bromo-o-toluonitrile
Formula: C₈H₆BrN
MW: 196.05 g/mol
Boiling Pt: 276 °C (1013 hPa)
Melting Pt: 70 °C
MDL Number: MFCD00041461
CAS Number: 67832-11-5
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR