4-Bromo-3-chlorobenzoic acid ≥97%

Warning

665312-5G 665312-25G
220086-409EA 47.71 USD
220086-409 220086-408
4-Bromo-3-chlorobenzoic acid ≥97%
4-Bromo-3-chlorobenzoic acid
Formula: C₇H₄BrClO₂
MW: 235.46 g/mol
MDL Number: MFCD08276864
CAS Number: 25118-59-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR