Methyl-5-iodo-2-methoxybenzoate ≥97%

Synonyms: Methyl 5-iodo-2-methoxybenzoate, 5-Iodo-2-methoxybenzoic acid methyl ester

665304-1G 665304-10G
220086-407EA 77.32 USD
220086-407 220086-406
Methyl-5-iodo-2-methoxybenzoate ≥97%
Methyl-5-iodo-2-methoxybenzoate
Formula: C₉H₉IO₃
MW: 292.07 g/mol
MDL Number: MFCD02683862
CAS Number: 40757-09-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR