5-Methoxyanthranilic acid ≥97%

Warning

665118-5G 665118-1G
220085-807EA 128.15 USD
220085-807 220085-806
5-Methoxyanthranilic acid ≥97%
5-Methoxyanthranilic acid
Formula: C₈H₉NO₃
MW: 167.16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00016509
CAS Number: 6705-03-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR