(4-Phenoxyphenyl)acetic acid ≥97%

Synonyms: 4-Phenoxyphenylacetic Acid

665053-1G 665053-5G
220086-951EA 47.38 USD
220086-951 220086-952
(4-Phenoxyphenyl)acetic acid ≥97%
(4-Phenoxyphenyl)acetic acid
Formula: (C₆H₅OC₆H₅)CH₂COOH
MW: 228.25 g/mol
MDL Number: MFCD00079779
CAS Number: 6328-74-1
UN: 3077
ADR: 9,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR