3-Chloro-1-phenyl-1-propyne ≥97%

Synonyms: (3-Chloroprop-1-yn-1-yl)benzene

664960-25G 664960-5G
220086-937EA 394.15 USD
220086-937 220086-938
3-Chloro-1-phenyl-1-propyne ≥97%
3-Chloro-1-phenyl-1-propyne
Formula: C₉H₇Cl
MW: 150.61 g/mol
MDL Number: MFCD06411085
CAS Number: 3355-31-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR