2-Bromo-5-chlorobenzoic acid ≥96%

664022-25G 664022-5G
220086-900EA 93.26 USD
220086-900 220086-901
2-Bromo-5-chlorobenzoic acid ≥96%
2-Bromo-5-chlorobenzoic acid
Formula: C₇H₄BrClO₂
MW: 235.46 g/mol
MDL Number: MFCD00013982
CAS Number: 21739-93-5
EINECS: 244-559-0
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR