4-Bromo-2-chlorobenzoic acid ≥97%

664014-5G 664014-25G
220086-899EA 40.92 USD
220086-899 220086-898
4-Bromo-2-chlorobenzoic acid ≥97%
4-Bromo-2-chlorobenzoic acid
Formula: C₇H₄BrClO₂
MW: 235.46 g/mol
MDL Number: MFCD00040903
CAS Number: 59748-90-2
EINECS: 261-911-9
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR