6-Fluoro-1-indanone

Warning

663123-5G 663123-1G
220086-752EA 207.69 USD
220086-752 220086-751
6-Fluoro-1-indanone
6-Fluoro-1-indanone
Formula: C₉H₇FO
MW: 150.15 g/mol
MDL Number: MFCD01318147
CAS Number: 1481-32-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR