5-Chloro-2-fluoropyridine ≥97%

Danger

662976-5G 662976-25G
220086-724EA 43.62 USD
220086-724 220086-723
5-Chloro-2-fluoropyridine ≥97%
5-Chloro-2-fluoropyridine
Formula: C₅H₃ClFN
MW: 131.54
Density: 1.311 g/mL (25 °C)
Flash Pt: 52 °C
MDL Number: MFCD04112513
CAS Number: 1480-65-5
UN: 1993
ADR: 3 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR