2-Hexynoic acid ≥97%

662895-25G 662895-5G
220086-376EA 445.85 USD
220086-376 220086-377
2-Hexynoic acid ≥97%
2-Hexynoic acid
Formula: C₆H₈O₂
MW: 112.13 g/mol
Boiling Pt: 110 - 112 °C (10 -rr)
Melting Pt: 23 - 26 °C
MDL Number: MFCD00014371
CAS Number: 764-33-0
UN: 3261
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR