3-Bromobutanone ≥97%

662747-1G 662747-5G
220086-362EA 92.29 USD
220086-362 220086-363
3-Bromobutanone ≥97%
3-Bromobutanone
Formula: H₃CC(O)BrCHCH₃
Boiling Pt: 36 °C (1013 hPa)
Density: 1.441 g/cm³ (18 °C)
MDL Number: MFCD00013538
CAS Number: 814-75-5
EINECS: 212-404-6
UN: 2920
ADR: 8,II

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR