4-Bromo-2-methylpyridine

662739-1G 662739-250MG
220086-360EA 60.74 USD
220086-360 220086-361
4-Bromo-2-methylpyridine
4-Bromo-2-methylpyridine
Formula: C₆H₆BrN
MW: 172.02 g/mol
MDL Number: MFCD03788196
CAS Number: 22282-99-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR