3-Bromo-2-methylpyridine

662720-1G 662720-250MG
220086-358EA 42 USD
220086-358 220086-359
3-Bromo-2-methylpyridine
3-Bromo-2-methylpyridine
Formula: C₆H₆BrN
MDL Number: MFCD00191224
CAS Number: 38749-79-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR