4-Amino-2-methylpyridine

Synonyms: 4-Amino-2-picoline, 2-Methyl-4-pyridylamine, 2-Methylpyridin-4-amine

662712-250MG 662712-1G
220086-357EA 88.31 USD
220086-357 220084-417
4-Amino-2-methylpyridine
4-Amino-2-methylpyridine
Formula: C₆H₈N₂
MW: 108.14 g/mol
MDL Number: MFCD00186509
CAS Number: 18437-58-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR