5-Amino-2-methylpyridine

Synonyms: 5-Amino-2-picoline, 6-Methylpyridin-3-amine, 3­Amino­6­methylpyridine, 6-Methyl-3-pyridinamine, 3­Amino­6­picoline

662704-1G 662704-250MG
220085-802EA 26.92 USD
220085-802 220085-803
5-Amino-2-methylpyridine
5-Amino-2-methylpyridine
Formula: C₆H₈N₂
MW: 108.14 g/mol
MDL Number: MFCD00833389
CAS Number: 3430-14-6
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR